Läs mer
Eseridine, TRC
Produktkod. 30411955
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411955 25 mg
30411758 250 mg
30409411 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30411955 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT12345025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 25573-43-7
Molekylformel C15 H21 N3 O3
Molekylvikt (g/mol) 291.35
Synonym 1,2-Oxazino[6,5-b]indol-6-ol, 2,3,4,4a,9,9a-hexahydro-2,4a,9-trimethyl-, methylcarbamate (ester), (4aS,9aS)- (9CI), 1,2-Oxazino[6,5-b]indol-6-ol, 2,3,4,4a,9,9a-hexahydro-2,4a,9-trimethyl-, methylcarbamate (ester), (4aS-cis)-, Geneserine (6CI,7CI,8CI), (-)-Geneserine, Eseridine, Eserine aminoxide, Eserine oxide, NSC 340071, Physostigmine aminoxide
IUPAC-namn [(4aS,9aS)-2,4a,9-trimethyl-4,9a-dihydro-3H-oxazino[6,5-b]indol-6-yl] N-methylcarbamate
LEDER CNC(=O)Oc1ccc2N(C)[C@H]3ON(C)CC[C@@]3(C)c2c1
Specifikationer
Kemiskt namn eller material Eseridine
Kvantitet 25 mg
InChI formel InChI=1S/C15H21N3O3/c1-15-7-8-17(3)21-13(15)18(4)12-6-5-10(9-11(12)15)20-14(19)16-2/h5-6,9,13H,7-8H2,1-4H3,(H,16,19)/t13-,15-/m0/s1
Formel vikt 291.1583