Läs mer
Tocris Bioscience™ ESI 09
Epac inhibitor
Brand: Tocris Bioscience™ 4773/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
ESI 09 is an EPAC inhibitor. Inhibits EPAC-mediated insulin releases from pancreatic β cells. Selective for EPAC over PKA. Decreases migration and invasion of AsPc-1 and PANC-1 pancreatic cancer cells in vitro.Specifikationer
| EPAC Inhibitors | |
| α-[(2-(3-Chlorophenyl)hydrazinylidene]-5-(1,1-dimethylethyl)-β-oxo-3-isoxazolepropanenitrile | |
| Soluble to 100mM in DMSO and to 20mM in ethanol | |
| Bioactive Small Molecule |
| 263707-16-0 | |
| 10 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use