Läs mer
Esmolol Dimer, TRC
Produktkod. 30373705
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30373705 100 mg
30373108 50 mg
30369223 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30373705 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE668030100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 98903-89-0
Molekylformel C31H46N2O7
Molekylvikt (g/mol) 558.71
Synonym 4-[2-Hydroxy-3-[[3-[4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]-1-oxopropyl](1-methylethyl)amino]propoxy]-benzenepropanoic Acid Methyl Ester
LEDER COC(=O)CCc1ccc(OCC(O)CN(C(C)C)C(=O)CCc2ccc(OCC(O)CNC(C)C)cc2)cc1
Specifikationer
Kemiskt namn eller material Esmolol Dimer
Kvantitet 100 mg
InChI formel InChI=1S/C31H46N2O7/c1-22(2)32-18-26(34)20-39-28-12-6-24(7-13-28)10-16-30(36)33(23(3)4)19-27(35)21-40-29-14-8-25(9-15-29)11-17-31(37)38-5/h6-9,12-15,22-23,26-27,32,34-35H,10-11,16-21H2,1-5H3
Formel vikt 558.3305