Läs mer
Esomeprazole Magnesium, TRC
Produktkod. 30372446
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30372446 100 mg
1 options
Produktkod. 30372446 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE668300100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Esomeprazole Magnesium Hydrate
CAS 668985-31-7
Molekylformel C34H36MgN6O6S2.xH2O
Kvantitet 100 mg
Synonym (S)-Omeprazole Magnesium Hydrate, (T-4)-Bis[6-methoxy-2-[(S)-[(4-Methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl-κO]-1H-benzimidazolato-κN3]-Magnesium Hydrate, Nexium Hydrate
InChI formel InChI=1S/2C17H18N3O3S.Mg.H2O/c2*1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17;;/h2*5-8H,9H2,1-4H3;;1H2/q2*-1;+2;
LEDER COC1=C(C(CS2=O[Mg+2]3(O=S(C4=[N]3C5=C([N-]4)C=C(OC)C=C5)CC6=NC=C(C(OC)=C6C)C)[N]7=C2[N-]C8=C7C=CC(OC)=C8)=NC=C1C)C.O
IUPAC-namn Esomeprazole Magnesium Hydrate
Molekylvikt (g/mol) 731.14
Formel vikt 730.21