missing translation for 'onlineSavingsMsg'
Learn More

ESR2 Mouse anti-Human, Mouse, Clone: 5D2C9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60197-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827195

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The human ER-beta is a newly discovered estrogen receptor initially cloned and characterized from testis. The size and structure of ER-beta is very similar to ER-alpha with the ligand and DNA binding domains being highly conserved, while the amino terminus which serves as their transactivation domain has diverged significantly. Similar in function to ER-alpha ER-beta binds to estrogen with a high affinity and regulates estrogen specific gene activation through direct interaction with estrogen response elements (ERE′s).
Specifications

Specifications

ESR2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08537, Q92731
ESR2
ESR2 Fusion Protein Ag5103
20 μL
Primary
Mouse, Human
Antibody
IgG1
Western Blot
5D2C9
Unconjugated
ESR2
ER BETA, Erb, ESR BETA, ESR2, ESRB, ESTRB, estrogen receptor 2 (ER beta), Estrogen receptor beta, NR3A2
Mouse
Protein G
RUO
13983, 2100
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers