Läs mer
Estradiol 17-Propionate, TRC
Produktkod. 30136026
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136026 25 mg
30006519 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136026 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE88881525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3758-34-7
Molekylformel C21H28O3
Molekylvikt (g/mol) 328.45
Synonym Estra-1,3,5(10)-triene-3,17-diol (17β)-, 17-propanoate, Estradiol, 17-propionate (7CI,8CI), 17β-Estradiol 17-propionate, Akrofollin, Estra-1,3,5(10)-triene-3,17β-diol 17-propionate, Estra-1,3,5(10)-triene-3,17β-diol 17β-propionate, Estradiol propionate, Oestra-1,3,5(10)-trien-3,17β-diol 17-propionate
IUPAC-namn [(8R,9S,13S,14S,17S)-3-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] propanoate
LEDER CCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Estradiol 17-Propionate
Kvantitet 25 mg
InChI formel InChI=1S/C21H28O3/c1-3-20(23)24-19-9-8-18-17-6-4-13-12-14(22)5-7-15(13)16(17)10-11-21(18,19)2/h5,7,12,16-19,22H,3-4,6,8-11H2,1-2H3/t16-,17-,18+,19+,21+/m1/s1
Formel vikt 328.2038