Läs mer
Estradiol 17-Valerate, TRC
Produktkod. 18003720
Change view
Kvantitet:
500 mg
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18003720 5 g
18000210 500 mg
18080600 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18003720 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE8888605G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 979-32-8
Molekylformel C23 H32 O3
Molekylvikt (g/mol) 356.5
Synonym Estradiol Valerate, (17β)-Estra-1,3,5(10)-triene-3,17-diol 17-pentanoate, Estra-1,3,5(10)-triene-3,17-diol (17β)-, 17-pentanoate (9CI, ACI), Estradiol valerate (6CI), Estradiol, 17-valerate (7CI, 8CI), 3-Hydroxy-17β-valeroyloxyestra-1,3,5(10)-triene, Atladiol, Climaval, Deladiol, Delahormone unimatic, Delestrogen, Delestrogen 4x, Dura-Estradiol, E2 valerate, Estra-1,3,5(10)-triene-3,17β-diol 17-valerate, Estradiol 17β-valerate, Estradiol valerat, Estradiol valerianate, Estraval, Femogex, Gynogen LA, Gynogen LA 40, Gynokadin, Meriestra, Neofollin, NSC 17590, Nuvelle, Oestradiol valerinate, Pelanin Depot, Pharlon, Primofol-Depot, Primogyn-Depot, Progynon-Depot, Progynova, Valergen, Valest
IUPAC-namn [(8R,9S,13S,14S,17S)-3-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] pentanoate
LEDER CCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material 17-beta-Estradiol-17-Valerate
Kvantitet 5 g
InChI formel InChI=1S/C23H32O3/c1-3-4-5-22(25)26-21-11-10-20-19-8-6-15-14-16(24)7-9-17(15)18(19)12-13-23(20,21)2/h7,9,14,18-21,24H,3-6,8,10-13H2,1-2H3/t18-,19-,20+,21+,23+/m1/s1
Formel vikt 356.2351