Learn More
Estradiol 3-Methyl Ether, Mikromol™
Product Code. 30051166
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051166 100 mg 100mg
30050838 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30051166 Supplier Mikromol Supplier No. MM0123.15
 Restricted Item
Beräknad leverans: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Estradiol 3-Methyl Ether
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Estradiol 3-Methyl Ether
CAS 1035-77-4
Analyte or Component Names Estradiol 3-Methyl Ether
Shipping Condition Room Temperature
Molecular Formula C19 H26 O2
Certifications/Compliance ISO 17025
Synonym Estra-1,3,5(10)-trien-17-ol, 3-methoxy-, (17β)-, Estra-1,3,5(10)-trien-17β-ol, 3-methoxy- (6CI,8CI), (17β)-3-Methoxyestra-1,3,5(10)-trien-17-ol, 17β-Estradiol 3-methyl ether, 17β-Hydroxy-3-methoxyestra-1,3,5(10)-triene, 3-Methoxy-13β-methyl-1,3,5(10)-gonatrien-17β-ol, 3-Methoxy-17β-hydroxyestra-1,3,5(10)-triene, 3-Methoxyestra-1,3,5(10)-trien-17β-ol, 3-Methoxyestra-1,3,5(10)-triene-17β-ol, 3-Methoxyestra-1,3,5(10)-triene-3,17β-diol, 3-Methoxyestradiol, 3-Methoxyoestra-1,3,5(10)-trien-17β-ol, 3-O-Methylestradiol, Estradiol 3-methyl ether, NSC 58851
InChI Formula InChI=1S/C19H26O2/c1-19-10-9-15-14-6-4-13(21-2)11-12(14)3-5-16(15)17(19)7-8-18(19)20/h4,6,11,15-18,20H,3,5,7-10H2,1-2H3/t15-,16-,17+,18+,19+/m1/s1
SMILES COc1ccc2[C@H]3CC[C@]4(C)[C@@H](O)CC[C@H]4[C@@H]3CCc2c1
IUPAC Name (8R,9S,13S,14S,17S)-3-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-ol
Molecular Weight (g/mol) 286.41
Quantity 100 mg
Formula Weight 286.1933 g/mol