Läs mer
Estradiol Benzoate, TRC
Produktkod. 30132910
Change view
Kvantitet:
50 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132910 1 g
30132664 500 mg
18013500 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132910 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE8882001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 50-50-0
Molekylformel C25 H28 O3
Molekylvikt (g/mol) 376.49
Synonym Estradiol Benzoate, (17β)-Estra-1,3,5(10)-triene-3,17-diol 3-benzoate, Estradiol 3-benzoate, 1,3,5(10)-Estratriene-3,17β-diol 3-benzoate, 17β-Estradiol 3-benzoate, 17β-Estradiol benzoate, 3-Benzoyloxy-17β-hydroxyestra-1,3,5(10)-triene, Agofollin, Agofollin Depot, Benovocylin, Benzhormovarine, Benzo-Gynoestryl, Benzoestrofol, Benzofoline, Benztrone, Cidirol, De Graafina, Diffollisterol, Difolliculine, Dimenformon benzoate, Diogyn B, Eston B, Estradiol benzoate, Estradiol monobenzoate, Estrogin, Femestrone, Folone, Graafina, Gynecormone, Gynformone, Hidroestron, Hormogynon, Mesalin, NSC 9566, Oestroform [BDH], Ovahormon benzoate, Ovasterol B, Ovocyclin Benzoate, Ovocyclin M, Ovocyclin MB, Pelanin benzoate, Primogyn B, Primogyn B oleosum, Progynon B, Progynon benzoate, Recthormone Oestradiol, Solestro, Unistradiol, β-Estradiol 3-benzoate, β-Estradiol benzoate
IUPAC-namn [(8R,9S,13S,14S,17S)-17-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl] benzoate
LEDER C[C@]12CC[C@H]3[C@@H](CCc4cc(OC(=O)c5ccccc5)ccc34)[C@@H]1CC[C@@H]2O
Specifikationer
Kemiskt namn eller material Estradiol Benzoate
Kvantitet 1 g
InChI formel InChI=1S/C25H28O3/c1-25-14-13-20-19-10-8-18(28-24(27)16-5-3-2-4-6-16)15-17(19)7-9-21(20)22(25)11-12-23(25)26/h2-6,8,10,15,20-23,26H,7,9,11-14H2,1H3/t20-,21-,22+,23+,25+/m1/s1
Formel vikt 376.2038