Läs mer
Estradiol Hemihydrate, Mikromol™
Beskrivning
Estradiol Hemihydrate
Specifikationer
Specifikationer
| Kemiskt namn eller material | Estradiol Hemihydrate |
| CAS | 35380-71-3 |
| Analyt- eller komponentnamn | Estradiol Hemihydrate |
| Frakt skick | Room Temperature |
| Molekylformel | 2 C18 H24 O2 . H2 O |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | Estradiol Benzoate Imp. A (EP), Estradiol Valerate Imp. A (EP), Estriol Imp. D (EP), Ethinylestradiol Imp. D (EP), Estradiol Hemihydrate, Estra-1,3,5(10)-triene-3,17-diol (17beta)-, hydrate (2:1) (9CI), beta-Estradiol hemihydrate, beta-Estradiol semihydrate, Estradiol Benzoate Imp. A (EP) as Hemihydrate, Estradiol Valerate Imp. A (EP) as Hemihydrate, Estriol Imp. D (EP) as Hemihydate, Ethinylestradiol Imp. D (EP) as Hemihydrate, Estradiol Benzoate Impurity A as Hemihydrate, Estradiol Valerate Impurity A as Hemihydrate, Estriol Impurity D as Hemihydate, Ethinylestradiol Impurity D as Hemihydrate |
| InChI formel | InChI=1S/2C18H24O2.H2O/c2*1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20;/h2*3,5,10,14-17,19-20H,2,4,6-9H2,1H3;1H2/t2*14-,15-,16+,17+,18+;/m11./s1 |
| LEDER | O.C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@H]2O.C[C@]56CC[C@H]7[C@@H](CCc8cc(O)ccc78)[C@@H]5CC[C@@H]6O |
| IUPAC-namn | (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol;hydrate |
| Visa mer |