missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Estradiol, MP Biomedicals™
Click to view available options
Quantity:
5 g
25 g
100 g
Specifications
Quantity | 5 g |
Formula Weight | 272.4 |
Physical Form | Solid |
Chemical Name or Material | β-Estradiol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'