Läs mer
Estradiol Valerate EP Impurity D, Mikromol™
Produktkod. 18012470
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18012470 25 mg
18011570 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18012470 Leverantör Mikromol Leverantörsnummer MM0209.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Estradiol Valerate EP Impurity D
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Methylestradiol Valerate
CAS 1359847-37-2
Analyt- eller komponentnamn 4-Methylestradiol Valerate
Frakt skick Room Temperature
Molekylformel C24 H34 O3
Certifikat och efterlevnad ISO 17025
Synonym 3-Hydroxy-4-methylestra-1,3,5(10)-trien-17β-yl Pentanoate (4-Methylestradiol Valerate), Estradiol Valerate Imp. D (EP), 3-Hydroxy-4-methylestra-1,3,5(10)-trien-17beta-yl Pentanoate, 4-Methylestradiol Valerate, Estradiol Valerate Impurity D
InChI formel InChI=1S/C13H19Cl/c1-9-6-11(13(3,4)5)7-10(2)12(9)8-14/h6-7H,8H2,1-5H5
LEDER CCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4c(C)c(O)ccc4[C@H]3CC[C@]12C
IUPAC-namn [(8R,9S,13S,14S,17S)-3-hydroxy-4,13-dimethyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] pentanoate
Molekylvikt (g/mol) 370.52
Kvantitet 25 mg
Formel vikt 370.2508 g/mol