Läs mer
Estradiol Valerate, LoGiCal
Produktkod. 30463961
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30463961 10 mg
1 options
Produktkod. 30463961 Leverantör LoGiCal Leverantörsnummer LGCFOR0209.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 17-beta-Estradiol-17-Valerate
CAS 979-32-8
Analyt- eller komponentnamn 17-beta-Estradiol-17-Valerate
Frakt skick Room Temperature
Molekylformel C23 H32 O3
Certifikat och efterlevnad ISO 9001
Synonym Estradiol Valerate, Estradiol 17-valerate, Atladiol, Climaval, Deladiol, Delahormone unimatic, Delestrogen, Delestrogen 4x, Dura-Estradiol, Estradiol valerianate, Estraval, Femogex, Gynogen LA, Gynogen LA 40, NSC 17590, Neofollin, Nuvelle, Oestradiol valerinate, Pelanin Depot, Pharlon, Primofol-Depot, Primogyn-Depot, Progynon-Depot, Progynova, Valergen, (17beta)-Estra-1,3,5(10)-triene-3,17-diol 17-pentanoate, 3-Hydroxy-17beta-valeroyloxyestra-1,3,5(10)-triene, Estra-1,3,5(10)-triene-3,17beta-diol 17-valerate, Estradiol 17beta-valerate
InChI formel InChI=1S/C23H32O3/c1-3-4-5-22(25)26-21-11-10-20-19-8-6-15-14-16(24)7-9-17(15)18(19)12-13-23(20,21)2/h7,9,14,18-21,24H,3-6,8,10-13H2,1-2H3/t18-,19-,20+,21+,23+/m1/s1
Lösningstyp Neat
LEDER CCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C
IUPAC-namn [(8R,9S,13S,14S,17S)-3-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] pentanoate
Industry Type Clinical Testing
Molekylvikt (g/mol) 356.5
Kvantitet 10 mg
Formel vikt 356.2351 g/mol