Läs mer
Estratetraenol, TRC
Produktkod. 30133189
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133189 10 mg
30135398 50 mg
30004322 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133189 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE88894010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1150-90-9
Molekylformel C18 H22 O
Molekylvikt (g/mol) 254.37
Synonym Estra-1,3,5(10),16-tetraen-3-ol, Compound 742
IUPAC-namn (8S,9S,13R,14S)-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-3-ol
LEDER C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC=C2
Specifikationer
Kemiskt namn eller material Estratetraenol
Kvantitet 10 mg
InChI formel InChI=1S/C18H22O/c1-18-9-2-3-17(18)16-6-4-12-11-13(19)5-7-14(12)15(16)8-10-18/h2,5,7,9,11,15-17,19H,3-4,6,8,10H2,1H3/t15-,16-,17+,18+/m1/s1
Formel vikt 254.1671