Läs mer
Estrone 3-Sulfate Sodium Salt, Stabilized with TRIS, TRC
Produktkod. 30375622
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30375622 100 mg
1 options
Produktkod. 30375622 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE889065100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Estrone Sodium Sulfate
CAS 438-67-5
Anteckningar om renhetsgrad HPLC
Molekylformel C18 H21 O5 S . Na
Kvantitet 100 mg
Synonym Estra-1,3,5(10)-trien-17-one, 3-(sulfooxy)-, sodium salt (1:1), Estra-1,3,5(10)-trien-17-one, 3-(sulfooxy)-, sodium salt (9CI), Estrone sulfate sodium salt (6CI), Estrone, hydrogen sulfate sodium salt (7CI,8CI), 3-Sulfatoxyestra-1,3,5(10)-trien-17-one sodium salt, Conestoral, Estrone sodium sulfate, Estrone sulfate sodium, Morestin, NSC 18313, Sodium estrone sulfate, Sodium estrone-3-sulfate
InChI formel InChI=1S/C18H22O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h3,5,10,14-16H,2,4,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t14-,15-,16+,18+;/m1./s1
LEDER [Na+].C[C@]12CC[C@H]3[C@@H](CCc4cc(OS(=O)(=O)[O-])ccc34)[C@@H]1CCC2=O
IUPAC-namn sodium;[(8R,9S,13S,14S)-13-methyl-17-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-yl] sulfate
Molekylvikt (g/mol) 372.41
Formel vikt 372.1007