Läs mer
Estropipate, Mikromol™
Beskrivning
Estropipate
Specifikationer
Specifikationer
| Kemiskt namn eller material | Estropipate |
| CAS | 7280-37-7 |
| Analyt- eller komponentnamn | Estropipate |
| Frakt skick | Room Temperature |
| Molekylformel | C18 H22 O5 S . C4 H10 N2 |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | Estra-1,3,5(10)-trien-17-one, 3-(sulfooxy)-, compd. with piperazine (1:1), Estrone, hydrogen sulfate, compd. with piperazine (1:1) (8CI), Piperazine, compd. with 3-(sulfooxy)estra-1,3,5(10)-trien-17-one (1:1) (9CI), Piperazine, compd. with estrone hydrogen sulfate (1:1) (8CI), 3-Sulfatoxyestra-1,3,5(10)-trien-17-one piperazine salt, Estrone sulfate piperazine salt, Estropipate, Harmogen, Ogen, Piperazine estrone sulfate, Sulestrex, Sulestrex piperazine |
| InChI formel | InChI=1S/C18H22O5S.C4H10N2/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;1-2-6-4-3-5-1/h3,5,10,14-16H,2,4,6-9H2,1H3,(H,20,21,22);5-6H,1-4H2/t14-,15-,16+,18+;/m1./s1 |
| LEDER | C[C@]12CC[C@H]3[C@@H](CCc4cc(OS(=O)(=O)O)ccc34)[C@@H]1CCC2=O.C5CNCCN5 |
| IUPAC-namn | [(8R,9S,13S,14S)-13-methyl-17-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-yl] hydrogen sulfate;piperazine |
| Visa mer |