Läs mer
Etafedrine Hydrochloride (mixture of diastereomers), TRC
Produktkod. 30136984
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136984 100 mg
30134639 25 mg
17944105 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136984 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE889200100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5591-29-7
Molekylformel C12 H19 N O . Cl H
Molekylvikt (g/mol) 229.75
Synonym Etafedrine Hydrochloride (mixture of diastereomers), Ethylephedrine Hydrochloride, α-[1-(Ethylmethylamino)ethyl]benzenemethanol Hydrochloride, α-[1-(Ethylmethylamino)ethyl]benzyl Alcohol Hydrochloride, Ditenate, Novedrine Hydrochloride
IUPAC-namn 2-[ethyl(methyl)amino]-1-phenylpropan-1-ol;hydrochloride
LEDER Cl.CCN(C)C(C)C(O)c1ccccc1
Specifikationer
Kemiskt namn eller material Etafedrine Hydrochloride (mixture of diastereomers)
Kvantitet 100 mg
InChI formel InChI=1S/C12H19NO.ClH/c1-4-13(3)10(2)12(14)11-8-6-5-7-9-11;/h5-10,12,14H,4H2,1-3H3;1H
Formel vikt 229.1233