missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETFA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ETFA |
---|---|
Concentration | 0.19 mg/mL |
Applications | Flow Cytometry, Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16839873
|
Proteintech
12262-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16829873
|
Proteintech
12262-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ETFA participates in catalyzing the initial step of the mitochondrial fatty acid beta-oxidation. It shuttles electrons between primary flavoprotein dehydrogenases and the membrane-bound electron transfer flavoprotein ubiquinone oxidoreductase. Defects in electron-transfer-flavoprotein have been implicated in type II glutaricaciduria in which multiple acyl-CoA dehydrogenase deficiencies result in large excretion of glutaric, lactic, ethylmalonic, butyric, isobutyric, 2-methyl-butyric, and isovaleric acids. Two transcript variants encoding different isoforms have been found for this gene.Specifications
ETFA | |
Flow Cytometry, Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P13803, P13804, Q99LC5 | |
110842, 2108, 300726 | |
ETFA Fusion Protein Ag2931 | |
Primary | |
-20°C | |
ETFA |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Alpha ETF, EMA, ETFA, GA2, MADD | |
ETFA | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |