Läs mer
Ethanethioic Acid S-[3-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)oxy]propyl] Ester, TRC
Produktkod. 30377553
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30377553 50 mg
1 options
Produktkod. 30377553 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE67646050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethanethioic Acid S-[3-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)oxy]propyl] Ester
CAS 1088-37-5
Molekylformel C13H13NO4S
Kvantitet 50 mg
Synonym Thioacetic Acid S-ester with N-(3-Mercaptopropoxy)phthalimide (7CI,8CI), NSC 101750
InChI formel InChI=1S/C13H13NO4S/c1-9(15)19-8-4-7-18-14-12(16)10-5-2-3-6-11(10)13(14)17/h2-3,5-6H,4,7-8H2,1H3
LEDER CC(=O)SCCCON1C(=O)c2ccccc2C1=O
IUPAC-namn S-[3-(1,3-dioxoisoindol-2-yl)oxypropyl] ethanethioate
Molekylvikt (g/mol) 279.31
Formel vikt 279.0565