missing translation for 'onlineSavingsMsg'
Learn More

Ethanol 70% + 100g IPA, Solveco

Catalog Number 11309206
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO

Prevents microbiological contamination. Solveco Ethanol 70% + 100g IPA provides a high degree of disinfection on smooth, visibly clean surfaces. It is effective against bacteria, fungi and viruses. The bottle can be used directly with a spray trigger.

Maintaining good hygiene in food production is crucial for avoiding infection/contamination of the product. Cleaning and disinfection should be performed regularly using Solveco Ethanol 70%, whose high level of disinfection has an important role in maintaining good microbiological quality of consumer products.

  • 70% ethanol by volume, denatured with isopropanol (100g/kg)
  • Clear, colorless, volatile liquid with a distinctive odor
  • Reacts explosively with strong oxidizing agents and can react violently with metals to produce hydrogen
  • May damage gaskets, painted surfaces, protective and sealing grease coatings, materials, rubber and some plastics
  • Molecular formula: CH3CH2OH
  • CAS: 64-17-5
Color Colorless
Formula Weight 46.07g/mol
Percent Purity 69.7–70.3%
Density 0.8847–0.8863kg/L
Quantity 1 L
Packaging HDPE Bottle
Water 6.5% max
compliance-icons compliance-icons
Product Identifier
  • Denaturerad Etanol 70% (denaturerad med Isopropanol)
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Etanol 70%, Propan-2-ol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'