missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, denaturated with 1% MEK and 0.001% Bitrex, 96%, Acros Organics™

Brand:  Acros Organics 457900050

Additional Details : CAS Number : 64-17-5 Weight : 3.95000kg Transport : UN number : 1170 Chem class : 1 Pack group : II

Quantity 5L

 View more versions of this product

Product Code. 15492182

  • 637.60 SEK / 5 L


Estimated Shipment date
from Supplier 01-03-2018
Add to basket

Specifications

Chemical Identifiers

0.7900g/mL
−114.0°C
46.06
12°C
C2H6O
Ethanol, denaturated with 1% MEK and 0.001% Bitrex
C2H5OH
1.2 mPa.s (20°C)
64-17-5
78.0°C
95.1 to 96.9% (calc. from rel. density)

Specifications

Ethanol, denaturated with 1% MEK and 0.001% Bitrex
95.1 to 96.9% (calc. from rel. density)
C2H5OH
0.7900g/mL
12°C
78.0°C
5L
3734-33-6
C2H6O
46.06
0.79
1.2 mPa.s (20°C)
−114.0°C
Videos