missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, denaturated with 1% MEK and 0.001% Bitrex, 96%, Acros Organics™

Brand:  Acros Organics 457900010

Additional Details : CAS Number : 64-17-5 Weight : 0.79000kg Transport : UN number : 1170 Chem class : 1 Pack group : II

Quantity 1L

 View more versions of this product

Product Code. 15482182

  • 215.38 SEK / 1 L


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

0.7900g/mL
−114.0°C
46.06
12°C
C2H6O
Ethanol, denaturated with 1% MEK and 0.001% Bitrex
C2H5OH
1.2 mPa.s (20°C)
64-17-5
78.0°C
96%

Specifications

Ethanol, denaturated with 1% MEK and 0.001% Bitrex
96%
C2H5OH
0.7900g/mL
12°C
78.0°C
1L
3734-33-6
C2H6O
46.06
0.79
1.2 mPa.s (20°C)
−114.0°C
Videos