missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, denatured, Spectrophotometric Grade, 90%, Alfa Aesar™

Brand:  Alfa Aesar 044014.K2

Additional Details : CAS Number : 64-17-5 Weight : 1.00000kg Transport : UN number : 1987 Chem class : 3 Pack group : II

Quantity 1L

 View more versions of this product

Product Code. 15434905

  • 693.33 SEK / 1 L


In stock, ready to ship: 1
Add to basket

Specifications

Chemical Identifiers

64-17-5
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol
C2H6O
MFCD00003568
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
CHEBI:16236
CCO

Specifications

Ethanol, denatured
Colorless liquid
C2H6O
1987
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
Miscible with water and most organic solvents.
CCO
46.069
CHEBI:16236
Liquid
0.789 g/cm3
Alcohol-like
1.361
11°C (52°F)
Colorless
1L
67-63-0
90%
MFCD00003568
1718733
Ethyl alcohol: 90.0%; Methanol: 5.0%; 2-Propanol: 5.0%
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
46.08
Spectrophotometric
455°C (851°F)
Resealable ChemSeal™ Bottles under Argon
57 hPa (43mm Hg) at 20°C
78°C
-90°C
Hygroscopic
Videos
SDS Documents