missing translation for 'onlineSavingsMsg'
Learn More

Ether, Anhydrous, HPLC Grade, J.T. Baker™

Catalog Number p-5383258
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
This item is not returnable. View return policy
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry, bulk pharmaceutical chemical
Flash Point −45.00°C (113°F)
Packaging Narrow Mouth Amber Glass Bottle
Quantity 4 L
Specific Gravity 1L = 0.71kg
Formula Weight 74.12
Grade Meets USP requirements
Chemical Name or Material Ether, Anhydrous
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'