missing translation for 'onlineSavingsMsg'
Learn More

Ethidium bromide, >98%, Molecular Biology Reagent, MP Biomedicals™

Catalog Number p-4589751
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1239-45-8
C21H20BrN3
394.32
MFCD00011724
ZMMJGEGLRURXTF-UHFFFAOYSA-N
ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
14710
CHEBI:4883
3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
This item is not returnable. View return policy
1239-45-8
C21H20BrN3
394.32
MFCD00011724
ZMMJGEGLRURXTF-UHFFFAOYSA-N
ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
14710
CHEBI:4883
3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Color Brown-Red
Assay Percent Range ≥98%
Quantity 1 g
Formula Weight 394.3
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material Ethidium bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'