missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethidium bromide, 99.11%, MP Biomedicals™
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Specifications
Color | Purple-Red |
Assay Percent Range | 1239-45-8 |
Quantity | 1 g |
Formula Weight | 394.3 |
Chemical Name or Material | Ethidium bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'