missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ethidium Bromide, Molecular Biology Grade, 98%
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Red to purple |
Formula Weight | 394.32 |
Melting Point | ca 261° dec. |
Odor | Weak |
InChI Key | ZMMJGEGLRURXTF-UHFFFAOYSA-N |
Product Type | Ethidium Bromide |
SMILES | [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12 |
IUPAC Name | 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide |
Molecular Weight (g/mol) | 394.32 |
Purity | Molecular Biology Grade |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'