missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Ethyl [1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetate (Indomethacin Ethyl Ester), Mikromol™
Beskrivning
Ethyl [1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetate (Indomethacin Ethyl Ester)
Specifikationer
Specifikationer
| Kemiskt namn eller material | Indomethacin Ethyl Ester |
| CAS | 16401-99-3 |
| Analyt- eller komponentnamn | Indomethacin Ethyl Ester |
| Frakt skick | Room Temperature |
| Molekylformel | C21 H20 Cl N O4 |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid ethyl ester, Indole-3-acetic acid, 1-(p-chlorobenzoyl)-5-methoxy-2-methyl-, ethyl ester (8CI), Ethyl 1-(4-chlorobenzoyl)-5-methoxy-2-methylindole-3-acetate, Ethyl 1-(p-chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetate, Indomethacin ethyl ester, Indometacin Impurity I |
| InChI formel | InChI=1S/C21H20ClNO4/c1-4-27-20(24)12-17-13(2)23(19-10-9-16(26-3)11-18(17)19)21(25)14-5-7-15(22)8-6-14/h5-11H,4,12H2,1-3H3 |
| LEDER | CCOC(=O)Cc1c(C)n(C(=O)c2ccc(Cl)cc2)c3ccc(OC)cc13 |
| IUPAC-namn | ethyl 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetate |
| Visa mer |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?