missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 1-Boc-piperazine-3-carboxylate, 97%

Brand:  Alfa Aesar™ H63146.06

Additional Details : CAS Number : 183742-29-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15449678

  • 5736.69 SEK / 5g


Estimated Shipment date
from Supplier 14-11-2018
Add to basket

Specifications

Chemical Identifiers

183742-29-2
258.318
IQKOVLZJPVVLOZ-UHFFFAOYSA-N
11402671
CCOC(=O)C1CN(CCN1)C(=O)OC(C)(C)C
C12H22N2O4
MFCD13172266
1-tert-butyl 3-ethyl piperazine-1,3-dicarboxylate, 1-boc-3-piperazinecarboxylic acid ethyl ester, 1-n-boc-3-piperazinecarboxylic acid ethyl ester, 1-tert-butoxycarbonyl-3-ethoxycarbonylpiperazine, 1-tert-butyl3-ethylpiperazine-1,3-dicarboxylate, 1-tert-butyl-3-ethylpiperazine-1,3-dicarboxylate, ethyl 4-tert-butoxycarbonyl-2-piperazinecarboxylate, 1-1,1-dimethylethyl 3-ethyl 1,3-piperazinedicarboxylate, 1-tert-butoxycarbonylpiperazine-3-carboxylic acid ethyl ester, ethyl 4-1,1-dimethylethoxycarbonyl-2-piperazinecarboxylate
1-O-tert-butyl 3-O-ethyl piperazine-1,3-dicarboxylate

Specifications

Ethyl 1-Boc-piperazine-3-carboxylate
183742-29-2
MFCD13172266
IQKOVLZJPVVLOZ-UHFFFAOYSA-N
1-O-tert-butyl 3-O-ethyl piperazine-1,3-dicarboxylate
11402671
Solid
5g
C12H22N2O4
1-tert-butyl 3-ethyl piperazine-1,3-dicarboxylate, 1-boc-3-piperazinecarboxylic acid ethyl ester, 1-n-boc-3-piperazinecarboxylic acid ethyl ester, 1-tert-butoxycarbonyl-3-ethoxycarbonylpiperazine, 1-tert-butyl3-ethylpiperazine-1,3-dicarboxylate, 1-tert-butyl-3-ethylpiperazine-1,3-dicarboxylate, ethyl 4-tert-butoxycarbonyl-2-piperazinecarboxylate, 1-1,1-dimethylethyl 3-ethyl 1,3-piperazinedicarboxylate, 1-tert-butoxycarbonylpiperazine-3-carboxylic acid ethyl ester, ethyl 4-1,1-dimethylethoxycarbonyl-2-piperazinecarboxylate
CCOC(=O)C1CN(CCN1)C(=O)OC(C)(C)C
258.318
258.31
97%
Videos
SDS