Läs mer
Ethyl 11,14-Diepoxyeicosanoate, TRC
Produktkod. 30132684
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30132684 2.5 mg 2.5mg
30133182 25 mg -
17998554 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132684 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9160002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 355803-78-0
Molekylformel C22 H40 O4
Molekylvikt (g/mol) 368.55
IUPAC-namn ethyl 10-[3-[(3-pentyloxiran-2-yl)methyl]oxiran-2-yl]decanoate
LEDER CCCCCC1OC1CC2OC2CCCCCCCCCC(=O)OCC
Specifikationer
Kemiskt namn eller material Ethyl 11,14-Diepoxyeicosanoate
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C22H40O4/c1-3-5-11-14-18-20(25-18)17-21-19(26-21)15-12-9-7-6-8-10-13-16-22(23)24-4-2/h18-21H,3-17H2,1-2H3
Formel vikt 368.2927
Procent renhet >95%