Läs mer
Ethyl 2-(1H-Pyrrol-2-yl)acetate, TRC
Produktkod. 30370891
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30370891 25 mg
1 options
Produktkod. 30370891 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92692825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2-(1H-Pyrrol-2-yl)acetate
CAS 4778-25-0
Molekylformel C8H11NO2
Kvantitet 25 mg
Synonym 1H-Pyrrole-2-acetic Acid Ethyl Ester, Pyrrole-2-acetic Acid Ethyl Ester, (1H-Pyrrol-2-yl)acetic Acid Ethyl Ester, 2-(Carboethoxymethyl)pyrrole, Ethyl Pyrrole-2-acetate, NSC 4510
InChI formel InChI=1S/C8H11NO2/c1-2-11-8(10)6-7-4-3-5-9-7/h3-5,9H,2,6H2,1H3
LEDER CCOC(=O)Cc1ccc[nH]1
IUPAC-namn ethyl 2-(1H-pyrrol-2-yl)acetate
Molekylvikt (g/mol) 153.18
Formel vikt 153.079