Läs mer
Ethyl 2,2,2-Trichloroacetate, TRC
Produktkod. 30377294
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30377294 5 g
1 options
Produktkod. 30377294 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE0783155G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trichloroacetate ethyl ester
CAS 515-84-4
Molekylformel C4 H5 Cl3 O2
Kvantitet 5 g
Synonym Trichloroacetic acid-ethyl ester, Acetic acid, 2,2,2-trichloro-, ethyl ester (ACI), Acetic acid, trichloro-, ethyl ester (6CI, 8CI, 9CI), Ethyl trichloroacetate, NSC 8829
InChI formel InChI=1S/C4H5Cl3O2/c1-2-9-3(8)4(5,6)7/h2H2,1H3
LEDER CCOC(=O)C(Cl)(Cl)Cl
IUPAC-namn ethyl 2,2,2-trichloroacetate
Molekylvikt (g/mol) 191.44
Formel vikt 189.9355