Läs mer
Ethyl 2-(2,6-Dichlorophenyl)acetate, TRC
Produktkod. 30386773
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30386773 250 mg
30374702 100 mg
30369361 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30386773 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE940928250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90793-64-9
Molekylformel C10H10O2Cl2
Molekylvikt (g/mol) 233.09
Synonym Ethyl 2,6-Dichlorobenzeneacetate, (2,6-Dichlorophenyl)acetic Acid Ethyl Ester
LEDER CCOC(=O)Cc1c(Cl)cccc1Cl
Specifikationer
Kemiskt namn eller material Ethyl 2-(2,6-Dichlorophenyl)acetate
Kvantitet 250 mg
InChI formel InChI=1S/C10H10Cl2O2/c1-2-14-10(13)6-7-8(11)4-3-5-9(7)12/h3-5H,2,6H2,1H3