Läs mer
Ethyl 2-(2-Chloro-4-pyrimidinyl)benzoate, TRC
Produktkod. 30377766
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30377766 50 mg
1 options
Produktkod. 30377766 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE86472050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2-(2-chloropyrimidin-4-yl)benzoate
CAS 954234-06-1
Molekylformel C13 H11 Cl N2 O2
Kvantitet 50 mg
Synonym GS-652118, Benzoic acid, 2-(2-chloro-4-pyrimidinyl)-, ethyl ester (ACI), Ethyl 2-(2-chloro-4-pyrimidinyl)benzoate (ACI)
InChI formel InChI=1S/C13H11ClN2O2/c1-2-18-12(17)10-6-4-3-5-9(10)11-7-8-15-13(14)16-11/h3-8H,2H2,1H3
LEDER CCOC(=O)c1ccccc1c2ccnc(Cl)n2
IUPAC-namn ethyl 2-(2-chloropyrimidin-4-yl)benzoate
Molekylvikt (g/mol) 262.692
Formel vikt 262.051