missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-(2-thienyl)thiazolidine-4-carboxylate, 97%

Brand:  Alfa Aesar™ H62388.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15448908

  • 8638.78 SEK / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-(2-thienyl)thiazolidine-4-carboxylate
243.35
MFCD26967916
2-(2-Thienyl)thiazolidine-4-carboxylic acid ethyl ester

Specifications

Ethyl 2-(2-thienyl)thiazolidine-4-carboxylate
5g
97%
MFCD26967916
Green
Liquid
243.35
2-(2-Thienyl)thiazolidine-4-carboxylic acid ethyl ester
Videos
SDS