Läs mer
Ethyl 2,3,4-Tri-O-acetyl-Beta-D-glucuronide, Methyl Ester, TRC
Produktkod. 30370023
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30370023 50 mg
1 options
Produktkod. 30370023 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92723550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2,3,4-Tri-O-acetyl-beta-D-glucuronide Methyl Ester
CAS 77392-66-6
Molekylformel C15 H22 O10
Kvantitet 50 mg
Synonym Ethyl 2,3,4-Tri-O-Acetyl-B-D-Glucuronide, Methyl Ester
InChI formel InChI=1S/C15H22O10/c1-6-21-15-13(24-9(4)18)11(23-8(3)17)10(22-7(2)16)12(25-15)14(19)20-5/h10-13,15H,6H2,1-5H3/t10-,11-,12-,13+,15+/m0/s1
LEDER CCO[C@@H]1O[C@@H]([C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]1OC(=O)C)C(=O)OC
IUPAC-namn methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-ethoxyoxane-2-carboxylate
Molekylvikt (g/mol) 362.33
Formel vikt 362.1213