Läs mer
Ethyl 2-(3-Cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate, TRC
Produktkod. 30133618
Change view
Kvantitet:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133618 5 g
30137783 1 g
30005776 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133618 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE9077805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 160844-75-7
Molekylformel C18 H20 N2 O3 S
Molekylvikt (g/mol) 344.43
Synonym 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid Ethyl Ester, Ethyl 2-(3-cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate
IUPAC-namn ethyl 2-[3-cyano-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylate
LEDER CCOC(=O)c1sc(nc1C)c2ccc(OCC(C)C)c(c2)C#N
Specifikationer
Kemiskt namn eller material 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid Ethyl Ester
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C18H20N2O3S/c1-5-22-18(21)16-12(4)20-17(24-16)13-6-7-15(14(8-13)9-19)23-10-11(2)3/h6-8,11H,5,10H2,1-4H3
Formel vikt 344.1195
Procent renhet >95%