Läs mer
Ethyl 2,3-Dicyanopropionate, TRC
Produktkod. 30378317
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30378317 500 mg
1 options
Produktkod. 30378317 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE915950500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2,3-Dicyanopropionate
CAS 40497-11-8
Molekylformel C7 H8 N2 O2
Kvantitet 500 mg
Synonym Propanoic acid, 2,3-dicyano-, ethyl ester, Propionic acid, 2,3-dicyano-, ethyl ester (6CI), 2,3-Dicyanopropanoic acid ethyl ester, 2,3-Dicyanopropionic acid ethyl ester, Ethyl 2,3-dicyanopropanoate, Ethyl 2,3-dicyanopropionate
InChI formel InChI=1S/C7H8N2O2/c1-2-11-7(10)6(5-9)3-4-8/h6H,2-3H2,1H3
LEDER CCOC(=O)C(CC#N)C#N
IUPAC-namn ethyl 2,3-dicyanopropanoate
Molekylvikt (g/mol) 152.15
Formel vikt 152.0586