Learn More
Alfa Aesar™ Ethyl 2-(3-pentyl)thiazolidine-4-carboxylate, 97%
Brand:  Alfa Aesar™ H62988.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15449418
  • 2477.86 SEK / 1g
Estimated Shipment date
from Supplier 22-10-2018
Add to basket
Specifications
Chemical Identifiers
MFCD26967921
2-(3-Pentyl)thiazolidine-4-carboxylic acid ethyl ester
Specifications
Ethyl 2-(3-pentyl)thiazolidine-4-carboxylate
1g
MFCD26967921
231.35
Colorless
97%
2-(3-Pentyl)thiazolidine-4-carboxylic acid ethyl ester