Läs mer
Ethyl 2-[[(3aR,4S,6R,6aS)-6-[(Benzyloxycarbonyl)amino]-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl]oxy]acetate, TRC
Produktkod. 30368820
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30368820 100 mg
1 options
Produktkod. 30368820 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE678775100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2-[((3aR,4S,6R,6aS)-6-[(Benzyloxycarbonyl)amino]-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl)oxy]acetate
CAS 866551-95-3
Molekylformel C20H27NO7
Kvantitet 100 mg
Synonym Ticagrelor Related Compound 49
InChI formel InChI=1S/C20H27NO7/c1-4-24-16(22)12-25-15-10-14(17-18(15)28-20(2,3)27-17)21-19(23)26-11-13-8-6-5-7-9-13/h5-9,14-15,17-18H,4,10-12H2,1-3H3,(H,21,23)/t14-,15+,17+,18-/m1/s1
LEDER O=C(OCC1=CC=CC=C1)N[C@H]2[C@@]3([H])[C@@](OC(C)(C)O3)([H])[C@@H](OCC(OCC)=O)C2
Molekylvikt (g/mol) 393.43
Formel vikt 393.1788