Läs mer
Ethyl 2-(4-Chlorophenoxy)carbanilate, TRC
Produktkod. 30137605
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137605 10 mg
30008939 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137605 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE90325010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 31879-60-4
Molekylformel C15H14ClNO3
Molekylvikt (g/mol) 291.73
LEDER CCOC(=O)Nc1ccccc1Oc2ccc(Cl)cc2
Specifikationer
Kemiskt namn eller material Ethyl 2-(4-Chlorophenoxy)carbanilate