Läs mer
Ethyl 2,4-Dichloro-5-sulphamoylbenzoate, Mikromol™
Produktkod. 30051505
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051505 100 mg
30051882 25 mg
30460479 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30051505 Leverantör Mikromol Leverantörsnummer MM0014.50
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 2,4-Dichloro-5-sulphamoylbenzoate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2,4-Dichloro-5-sulphamoylbenzoate
CAS 4793-25-3
Analyt- eller komponentnamn Ethyl 2,4-Dichloro-5-sulphamoylbenzoate
Frakt skick Room Temperature
Molekylformel C9 H9 Cl2 N O4 S
Certifikat och efterlevnad ISO 17025
Synonym Ethyl 2,4-Dichloro-5-sulphamoylbenzoate
InChI formel InChI=1S/C9H9Cl2NO4S/c1-2-16-9(13)5-3-8(17(12,14)15)7(11)4-6(5)10/h3-4H,2H2,1H3,(H2,12,14,15)
LEDER CCOC(=O)c1cc(c(Cl)cc1Cl)S(=O)(=O)N
IUPAC-namn ethyl 2,4-dichloro-5-sulfamoylbenzoate
Molekylvikt (g/mol) 298.14
Kvantitet 100 mg
Formel vikt 296.9629 g/mol