missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-dichloropyrimidine-5-carboxylate, 98%, Thermo Scientific Chemicals

Catalog Number p-7056046
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
51940-64-8
C7H6Cl2N2O2
221.037
MFCD09910281
SRJBDGLSCPDXBL-UHFFFAOYSA-N
104020
ethyl 2,4-dichloropyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1Cl)Cl
This item is not returnable. View return policy
51940-64-8
C7H6Cl2N2O2
221.037
MFCD09910281
SRJBDGLSCPDXBL-UHFFFAOYSA-N
104020
ethyl 2,4-dichloropyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1Cl)Cl

It's employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 36°C to 37°C
Color White
Quantity 250 mg
Beilstein 152173
Synonym ethyl 2,4-dichloro-5-pyrimidinecarboxylate, 5-pyrimidinecarboxylic acid, 2,4-dichloro-, ethyl ester, ethyl 2,4-dichloro-pyrimidine-5-carboxylate, 2,4-dichloro-pyrimidine-5-carboxylic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonylpyrimidine, 2,4,-dichloro-5-pyrimidinecarboxylic acid ethyl ester, 2,4-dichloro-5-pyrimidinecarboxylic acid ethyl ester, 2,4-dichloropyrimidine-5-carboxylic acid ethyl ester, pubchem24010, ethyl2,4-dichloro-5-pyrimidinecarboxylate
Solubility Information Slightly soluble in water.
Formula Weight 221.04
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Ethyl 2,4-dichloropyrimidine-5-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'