missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-dioxovalerate, 98%, Thermo Scientific Chemicals

Catalog Number p-3757853
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
615-79-2
C7H10O4
158.15
MFCD00009124
OYQVQWIASIXXRT-UHFFFAOYSA-N
ethyl 2,4-dioxovalerate, ethyl acetopyruvate, pentanoic acid, 2,4-dioxo-, ethyl ester, ethyl2,4-dioxovalerate, 2,4-dioxo-pentanoic acid ethyl ester, 2,4-dioxopentanoic acid ethyl ester, 2,4-dioxo-pentanoic-acid-ethyl-ester, 2,4-dioxo-pentanoicacid ethylester, ethyl 2,4-dioxo-valerate, ethyl 2,4-dioxo-pentanoate
69208
ethyl 2,4-dioxopentanoate
CCOC(=O)C(=O)CC(C)=O
This item is not returnable. View return policy
615-79-2
C7H10O4
158.15
MFCD00009124
OYQVQWIASIXXRT-UHFFFAOYSA-N
ethyl 2,4-dioxovalerate, ethyl acetopyruvate, pentanoic acid, 2,4-dioxo-, ethyl ester, ethyl2,4-dioxovalerate, 2,4-dioxo-pentanoic acid ethyl ester, 2,4-dioxopentanoic acid ethyl ester, 2,4-dioxo-pentanoic-acid-ethyl-ester, 2,4-dioxo-pentanoicacid ethylester, ethyl 2,4-dioxo-valerate, ethyl 2,4-dioxo-pentanoate
69208
ethyl 2,4-dioxopentanoate
CCOC(=O)C(=O)CC(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 16.0°C to 18.0°C
Color Yellow
Density 1.1200g/mL
Boiling Point 101.0°C to 103.0°C (12.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH2COCO2C2H5
Refractive Index 1.4730 to 1.4750
Quantity 5 g
Beilstein 03, 747
Specific Gravity 1.12
Formula Weight 158.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 2,4-dioxovalerate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'