Läs mer
Ethyl 2-Acetamido-2-deoxy-Beta-D-thioglucopyranoside, TRC
Produktkod. 30372765
Change view
Kvantitet:
200 mg
Produktkod. Kvantitet
30372765 200 mg
1 options
Produktkod. 30372765 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE894750200MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2-Acetamido-2-deoxy-beta-D-thioglucopyranoside
CAS 122331-70-8
Molekylformel C10H19NO5S
Kvantitet 200 mg
Synonym Ethyl 2-(Acetylamino)-2-deoxy-1-thio-beta-D-glucopyranoside
InChI formel InChI=1S/C10H19NO5S/c1-3-17-10-7(11-5(2)13)9(15)8(14)6(4-12)16-10/h6-10,12,14-15H,3-4H2,1-2H3,(H,11,13)/t6-,7-,8-,9-,10+/m1/s1
LEDER CCS[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(=O)C
Molekylvikt (g/mol) 265.33
Formel vikt 265.0984