missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-acetylisonicotinate, 96%

Brand:  Alfa Aesar™ H36911.03

Additional Details : CAS Number : 25028-32-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15437717

  • 787.32 SEK / 1g


Estimated Shipment
30-09-2020
Add to basket

Specifications

Chemical Identifiers

25028-32-4
193.202
LUMZQFHOBXCXOR-UHFFFAOYSA-N
55265486
CCOC(=O)C1=CC(=NC=C1)C(=O)C
C10H11NO3
MFCD11042531
ethyl 2-acetylisonicotinate, 2-acetyl-4-pyridinecarboxylic acid ethyl ester, pyac32, 2-acetyl-isonicotinic acid ethyl ester, 2-acetylpyridine-4-carboxylic acid ethyl ester
ethyl 2-acetylpyridine-4-carboxylate

Specifications

Ethyl 2-acetylisonicotinate
52°C to 55°C
25028-32-4
MFCD11042531
LUMZQFHOBXCXOR-UHFFFAOYSA-N
ethyl 2-acetylpyridine-4-carboxylate
55265486
Fused Solid
Cream
1g
C10H11NO3
ethyl 2-acetylisonicotinate, 2-acetyl-4-pyridinecarboxylic acid ethyl ester, pyac32, 2-acetyl-isonicotinic acid ethyl ester, 2-acetylpyridine-4-carboxylic acid ethyl ester
CCOC(=O)C1=CC(=NC=C1)C(=O)C
193.202
193.2
96%
Videos
SDS Documents