missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, 97%, Thermo Scientific™

Catalog Number 15206018
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
4815-24-1
C9H13NO2S
199.27
JYSDXODDWAQWJR-UHFFFAOYSA-N
ethyl 2-amino-4,5-dimethyl3-thenoate, ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, 2-amino-4,5-dimethylthiophene-3-carboxylic acid ethyl ester, 2-amino-4,5-dimethyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4,5-dimethyl-, ethyl ester, 2-amino-3-carbethoxy-4,5-dimethylthiophene, 2-amino-4,5-dimethyl-3-ethoxycarbonyl-thiophene, ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl 2-amino-4,5-dimethyl-thiophene-3-carboxylate
78536
ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
CCOC(=O)C1=C(SC(=C1C)C)N
This item is not returnable. View return policy
4815-24-1
C9H13NO2S
199.27
JYSDXODDWAQWJR-UHFFFAOYSA-N
ethyl 2-amino-4,5-dimethyl3-thenoate, ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, 2-amino-4,5-dimethylthiophene-3-carboxylic acid ethyl ester, 2-amino-4,5-dimethyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4,5-dimethyl-, ethyl ester, 2-amino-3-carbethoxy-4,5-dimethylthiophene, 2-amino-4,5-dimethyl-3-ethoxycarbonyl-thiophene, ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl 2-amino-4,5-dimethyl-thiophene-3-carboxylate
78536
ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
CCOC(=O)C1=C(SC(=C1C)C)N
Melting Point 90.0°C to 91.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 10 g
Formula Weight 199.27
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate
compliance-icons
Product Identifier
  • Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'