missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-biphenylylthiophene-3-carboxylate, 97%, Thermo Scientific™

Catalog Number p-7039658
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
307343-50-6
C19H17NO2S
323.41
MFCD01912029
GGRXYEXAHRYAIO-UHFFFAOYSA-N
ethyl 2-amino-4-1,1'-biphenyl-4-yl-3-thiophenecarboxylate, ethyl 2-amino-4-4-phenylphenyl thiophene-3-carboxylate, ethyl 2-amino-4-1,1-biphenyl-4-yl-3-thiophenecarboxylate, ethyl 2-amino-4-1,1'-biphenyl-4-yl thiophene-3-carboxylate, ethyl 2-amino-4-biphenyl-4-yl thiophene-3-carboxylate, 3-thiophenecarboxylicacid, 2-amino-4-1,1'-biphenyl-4-yl-, ethyl ester, cbmicro_001199, ethyl 2-amino-4-biphenylylthiophene-3-carboxylate
722488
ethyl 2-amino-4-(4-phenylphenyl)thiophene-3-carboxylate
CCOC(=O)C1=C(N)SC=C1C1=CC=C(C=C1)C1=CC=CC=C1
This item is not returnable. View return policy
307343-50-6
C19H17NO2S
323.41
MFCD01912029
GGRXYEXAHRYAIO-UHFFFAOYSA-N
ethyl 2-amino-4-1,1'-biphenyl-4-yl-3-thiophenecarboxylate, ethyl 2-amino-4-4-phenylphenyl thiophene-3-carboxylate, ethyl 2-amino-4-1,1-biphenyl-4-yl-3-thiophenecarboxylate, ethyl 2-amino-4-1,1'-biphenyl-4-yl thiophene-3-carboxylate, ethyl 2-amino-4-biphenyl-4-yl thiophene-3-carboxylate, 3-thiophenecarboxylicacid, 2-amino-4-1,1'-biphenyl-4-yl-, ethyl ester, cbmicro_001199, ethyl 2-amino-4-biphenylylthiophene-3-carboxylate
722488
ethyl 2-amino-4-(4-phenylphenyl)thiophene-3-carboxylate
CCOC(=O)C1=C(N)SC=C1C1=CC=C(C=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 323.41
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-biphenylylthiophene-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'