Learn More
Alfa Aesar™ Ethyl 2-aminothiophene-3-carboxylate, 98%
1222.81 SEK - 4419.27 SEK
Chemical Identifiers
CAS 31891-06-2
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD01566303
InChI Key MKJQYFVTEPGXIE-UHFFFAOYSA-N
Synonym 2-amino-thiophene-3-carboxylic acid ethyl ester, 2-aminothiophene-3-carboxylic acid ethyl ester, ethyl2-aminothiophene-3-carboxylate, 2-amino-3-ethoxycarbonylthiophene, ethyl 2-amino-3-thiophenecarboxylate, 2-aminothiophene-3-carboxylate, 3-thiophenecarboxylic acid, 2-amino-, ethyl ester, pubchem22914, acmc-209hpg, ethyl 2-amino-3-thenoate
PubChem CID 1988156
IUPAC Name ethyl 2-aminothiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1)N
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15428128
SDS View Documents
Alfa Aesar™
H61559.14
25g 1222.81 SEK
25g View alternative products
Save Up to 
»
In stock, ready to ship: 1
Add to basket
15438128
SDS View Documents
Alfa Aesar™
H61559.22
100g 4419.27 SEK
100g View alternative products
Save Up to 
»
Estimated Shipment
17-11-2020
Add to basket
Description
Description
Applications
Ethyl 2-aminothiophene-3-carboxylate is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Chemical Identifiers
31891-06-2
171.214
MKJQYFVTEPGXIE-UHFFFAOYSA-N
1988156
CCOC(=O)C1=C(SC=C1)N
C7H9NO2S
MFCD01566303
2-amino-thiophene-3-carboxylic acid ethyl ester, 2-aminothiophene-3-carboxylic acid ethyl ester, ethyl2-aminothiophene-3-carboxylate, 2-amino-3-ethoxycarbonylthiophene, ethyl 2-amino-3-thiophenecarboxylate, 2-aminothiophene-3-carboxylate, 3-thiophenecarboxylic acid, 2-amino-, ethyl ester, pubchem22914, acmc-209hpg, ethyl 2-amino-3-thenoate
ethyl 2-aminothiophene-3-carboxylate
Specifications
Ethyl 2-aminothiophene-3-carboxylate
31891-06-2
MFCD01566303
Slightly soluble in water.
CCOC(=O)C1=C(SC=C1)N
171.214
171.22
98%
25g
C7H9NO2S
2-amino-thiophene-3-carboxylic acid ethyl ester, 2-aminothiophene-3-carboxylic acid ethyl ester, ethyl2-aminothiophene-3-carboxylate, 2-amino-3-ethoxycarbonylthiophene, ethyl 2-amino-3-thiophenecarboxylate, 2-aminothiophene-3-carboxylate, 3-thiophenecarboxylic acid, 2-amino-, ethyl ester, pubchem22914, acmc-209hpg, ethyl 2-amino-3-thenoate
MKJQYFVTEPGXIE-UHFFFAOYSA-N
ethyl 2-aminothiophene-3-carboxylate
1988156
Solid