missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-bromothiazole-5-carboxylate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Ethyl 2-bromothiazole-5-carboxylate is used in medicine, pharmaceutical intermediate
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 2-bromothiazole-5-carboxylate is used in medicine, pharmaceutical intermediate
Solubility
Miscible is water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Ethyl 2-bromothiazole-5-carboxylate is used in medicine, pharmaceutical intermediate
Solubility
Miscible is water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Density | 1.581 |
Boiling Point | 216°C to 217°C |
Flash Point | 110°C (230°F) |
Refractive Index | 1.553 |
Quantity | 250 mg |
Synonym | ethyl 2-bromothiazole-5-carboxylate, 2-bromo-thiazole-5-carboxylic acid ethyl ester, ethyl 2-bromo-5-thiazolecarboxylate, 2-bromothiazole-5-carboxylic acid ethyl ester, 2-bromo-5-ethoxycarbonyl thiazole, ethyl-2-bromothiazole-5-carboxylate, ethyl2-bromo-5-thiazolecarboxylate, ethyl2-bromothiazole-5-carboxylate, pubchem13480, acmc-1ambg |
Solubility Information | Miscible is water. |
Formula Weight | 236.08 |
Percent Purity | 98% |
Chemical Name or Material | Ethyl 2-bromothiazole-5-carboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'